Merck
  • Novel mammalian cell cycle inhibitors, tryprostatins A, B and other diketopiperazines produced by Aspergillus fumigatus. I. Taxonomy, fermentation, isolation and biological properties.
Novel mammalian cell cycle inhibitors, tryprostatins A, B and other diketopiperazines produced by Aspergillus fumigatus. I. Taxonomy, fermentation, isolation and biological properties.
The Journal of antibiotics (1996-06-01)
C B Cui, H Kakeya, G Okada, R Onose, H Osada
ABSTRACT
Two novel diketopiperazines named tryprostatins A (1) and B (2) and a new natural product belonging to the diketopiperazine series, designated as demethoxyfumitremorgin C (3), together with four known diketopiperazines, fumitremorgin C (4), 12,13-dihydroxyfumitremorgin C (5), fumitremorgin B (6) and verruculogen (7), were isolated from the fermentation broth of Aspergillus fumigatus BM939 by the combined use of solvent extraction, silica gel column chromatography, preparative TLC and repeated-preparative HPLC. The diketopiperazines showed an inhibitory activity on the cell cycle progression of mouse tsFT210 cells in the M phase with the MIC values of 16.4 microM (1), 4.4 microM (2), 0.45 microM (3), 4.1 microM (4), 60.8 microM (5), 26.1 microM (6) and 12.2 microM (7), respectively.