Merck
  • Human β-defensin-3 attenuates atopic dermatitis-like inflammation through autophagy activation and the aryl hydrocarbon receptor signaling pathway.
Human β-defensin-3 attenuates atopic dermatitis-like inflammation through autophagy activation and the aryl hydrocarbon receptor signaling pathway.
The Journal of clinical investigation (2022-07-15)
Ge Peng, Saya Tsukamoto, Risa Ikutama, Hai Le Thanh Nguyen, Yoshie Umehara, Juan V Trujillo-Paez, Hainan Yue, Miho Takahashi, Takasuke Ogawa, Ryoma Kishi, Mitsutoshi Tominaga, Kenji Takamori, Jiro Kitaura, Shun Kageyama, Masaaki Komatsu, Ko Okumura, Hideoki Ogawa, Shigaku Ikeda, François Niyonsaba
ABSTRACT
Human β-defensin-3 (hBD-3) exhibits antimicrobial and immunomodulatory activities; however, its contribution to autophagy regulation remains unclear, and the role of autophagy in the regulation of the epidermal barrier in atopic dermatitis (AD) is poorly understood. Here, keratinocyte autophagy was restrained in the skin lesions of patients with AD and murine models of AD. Interestingly, hBD-3 alleviated the IL-4- and IL-13-mediated impairment of the tight junction (TJ) barrier through keratinocyte autophagy activation, which involved aryl hydrocarbon receptor (AhR) signaling. While autophagy deficiency impaired the epidermal barrier and exacerbated inflammation, hBD-3 attenuated skin inflammation and enhanced the TJ barrier in AD. Importantly, hBD-3-mediated improvement of the TJ barrier was abolished in autophagy-deficient AD mice and in AhR-suppressed AD mice, suggesting a role for hBD-3-mediated autophagy in the regulation of the epidermal barrier and inflammation in AD. Thus, autophagy contributes to the pathogenesis of AD, and hBD-3 could be used for therapeutic purposes.