Merck
  • Stable adducts of nerve agents sarin, soman and cyclosarin with TRIS, TES and related buffer compounds--characterization by LC-ESI-MS/MS and NMR and implications for analytical chemistry.
Stable adducts of nerve agents sarin, soman and cyclosarin with TRIS, TES and related buffer compounds--characterization by LC-ESI-MS/MS and NMR and implications for analytical chemistry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-02-23)
Jürgen Gäb, Harald John, Marco Melzer, Marc-Michael Blum
ABSTRACT
Buffering compounds like TRIS are frequently used in chemical, biochemical and biomedical applications to control pH in solution. One of the prerequisites of a buffer compound, in addition to sufficient buffering capacity and pH stability over time, is its non-reactivity with other constituents of the solution. This is especially important in the field of analytical chemistry where analytes are to be determined quantitatively. Investigating the enzymatic hydrolysis of G-type nerve agents sarin, soman and cyclosarin in buffered solution we have identified stable buffer adducts of TRIS, TES and other buffer compounds with the nerve agents. We identified the molecular structure of these adducts as phosphonic diesters using 1D (1)H-(31)P HSQC NMR and LC-ESI-MS/MS techniques. Reaction rates with TRIS and TES are fast enough to compete with spontaneous hydrolysis in aqueous solution and to yield substantial amounts (up to 20-40%) of buffer adduct over the course of several hours. A reaction mechanism is proposed in which the amino function of the buffer serves as an intramolecular proton acceptor rendering the buffer hydroxyl groups nucleophilic enough for attack on the phosphorus atom of the agents. Results show that similar buffer adducts are formed with a range of hydroxyl and amino function containing buffers including TES, BES, TRIS, BIS-TRIS, BIS-TRIS propane, Tricine, Bicine, HEPES and triethanol amine. It is recommended to use alternative buffers like MOPS, MES and CHES when working with G-type nerve agents especially at higher concentrations and over prolonged times.
Sigma-Aldrich
TES, ≥99% (titration)
Sigma-Aldrich
TES, Vetec, reagent grade, 99%