Merck
  • Trifluoroethanol and acetonitrile induced formation of the molten globule states and aggregates of cellulase.
Trifluoroethanol and acetonitrile induced formation of the molten globule states and aggregates of cellulase.
International journal of biological macromolecules (2012-06-09)
Afshin Iram, Aabgeena Naeem
ABSTRACT
A systematic investigation on the effects of trifluoroethanol and acetonitrile at various concentrations on cellulase (EC 3.2.1.4) was studied by enzyme assay, intrinsic fluorescence, ANS binding, circular dichroism and ATR-Fourier transform infra red spectroscopy. The results show the presence of molten globule states at 3% (v/v) TFE and 80% (v/v) ACN. Cellulase aggregates at 25% (v/v) TFE and 90% (v/v) ACN, as detected by decrease in intrinsic and ANS fluorescence, loss in tertiary structure and enzyme activity, increase non-native β-sheet structure as evident from far-UV CD and FTIR spectra, increase in Thioflavin T fluorescence and shift in Congo red assay.
Sigma-Aldrich
2,2,2-Trifluoroethanol, ReagentPlus®, ≥99%
Supelco
2,2,2-Trifluoroethanol, analytical standard, suitable for NMR (spectroscopy)