Merck
  • Synthesis and biological activity of artificial mRNA prepared with novel phosphorylating reagents.
Synthesis and biological activity of artificial mRNA prepared with novel phosphorylating reagents.
Nucleic acids research (2010-07-28)
Seigo Nagata, Tomohiro Hamasaki, Koichi Uetake, Hirofumi Masuda, Kazuchika Takagaki, Natsuhisa Oka, Takeshi Wada, Tadaaki Ohgi, Junichi Yano
ABSTRACT
Though medicines that target mRNA are under active investigation, there has been little or no effort to develop mRNA itself as a medicine. Here, we report the synthesis of a 130-nt mRNA sequence encoding a 33-amino-acid peptide that includes the sequence of glucagon-like peptide-1, a peptide that stimulates glucose-dependent insulin secretion from the pancreas. The synthesis method used, which had previously been developed in our laboratory, was based on the use of 2-cyanoethoxymethyl as the 2'-hydroxy protecting group. We also developed novel, highly reactive phosphotriester pyrophosphorylating reagents to pyrophosphorylate the 5'-end of the 130-mer RNA in preparation for capping. We completed the synthesis of the artificial mRNA by the enzymatic addition of a 5'-cap and a 3'-poly(A) tail to the pyrophosphorylated 130-mer and showed that the resulting mRNA supported protein synthesis in a cell-free system and in whole cells. As far as we know, this is the first time that mRNA has been prepared from a chemically synthesized RNA sequence. As well as providing a research tool for the intracellular expression of peptides, the technology described here may be used for the production of mRNA for medical applications.
Sigma-Aldrich
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite, 95%
Pricing and availability is not currently available.