Merck
  • ABT-702 (4-amino-5-(3-bromophenyl)-7-(6-morpholinopyridin-3-yl)pyrido[2, 3-d]pyrimidine), a novel orally effective adenosine kinase inhibitor with analgesic and anti-inflammatory properties: I. In vitro characterization and acute antinociceptive effects in the mouse.
ABT-702 (4-amino-5-(3-bromophenyl)-7-(6-morpholinopyridin-3-yl)pyrido[2, 3-d]pyrimidine), a novel orally effective adenosine kinase inhibitor with analgesic and anti-inflammatory properties: I. In vitro characterization and acute antinociceptive effects in the mouse.
The Journal of pharmacology and experimental therapeutics (2000-11-18)
M F Jarvis, H Yu, K Kohlhaas, K Alexander, C H Lee, M Jiang, S S Bhagwat, M Williams, E A Kowaluk
ABSTRACT
Adenosine (ADO) is an inhibitory neuromodulator that can increase nociceptive thresholds in response to noxious stimulation. Inhibition of the ADO-metabolizing enzyme adenosine kinase (AK) increases extracellular ADO concentrations at sites of tissue trauma and AK inhibitors may have therapeutic potential as analgesic and anti-inflammatory agents. ABT-702 is a novel and potent (IC(50) = 1. 7 nM) non-nucleoside AK inhibitor that has several orders of magnitude selectivity over other sites of ADO interaction (A(1), A(2A), A(3) receptors, ADO transporter, and ADO deaminase). ABT-702 was 1300- to 7700-fold selective for AK compared with a number of other neurotransmitter and peptide receptors, ion channel proteins, neurotransmitter/nucleoside reuptake sites, and enzymes, including cycloxygenases-1 and -2. ABT-702 was equipotent (IC(50) = 1.5 +/- 0. 3 nM) in inhibiting native human AK (placenta), two human recombinant isoforms (AK(long) and AK(short)), and AK from monkey, dog, rat, and mouse brain. Kinetic studies revealed that AK inhibition by ABT-702 was competitive with respect to ADO and noncompetitive with respect to MgATP(2-). AK inhibition by ABT-702 was demonstrated to be reversible after 4 h of dialysis. ABT-702 is orally active and fully efficacious in reducing acute somatic nociception (ED(50) = 8 micromol/kg i.p.; 65 micromol/kg p.o.) in the mouse hot-plate assay. ABT-702 also dose dependently reduced nociception in the phenyl-p-quinone-induced abdominal constriction assay. The antinociceptive effects of ABT-702 in the hot-plate assay were blocked by the nonselective ADO receptor antagonist theophylline, and by the A(1)-selective antagonist cyclopentyltheophylline (10 mg/kg i.p.), but not by a peripherally selective ADO receptor antagonist 8-(p-sulfophenyl)-theophylline (50 mg/kg i.p.), by the A(2A)-selective antagonist 3, 7-dimethyl-1-propargylxanthine (1 mg/kg i.p.) or the opioid antagonist naloxone (5 mg/kg i.p.). Thus, ABT-702 is a novel and potent non-nucleoside AK inhibitor that effectively reduces acute thermal nociception in the mouse by a nonopioid, non-nonsteroidal anti-inflammatory drug, ADO A(1) receptor-mediated mechanism.
Product Description
Millipore
Fractogel® EMD SO3⁻ (S) Resin, strong cation exchanger, suspension in 20% ethanol and 150 mM NaCl (20-40 µm)