Merck
  • Hydrodynamic transformation of a freestanding polymer nanosheet induced by a thermoresponsive surface.
Hydrodynamic transformation of a freestanding polymer nanosheet induced by a thermoresponsive surface.
ACS applied materials & interfaces (2010-04-02)
Toshinori Fujie, Jin Young Park, Atsushi Murata, Nicel C Estillore, Maria Celeste R Tria, Shinji Takeoka, Rigoberto C Advincula
ABSTRACT
Freestanding quasi-two-dimensional ultrathin films (e.g., 41 nm thick polymer nanosheets) were produced, on which stimuli-responsive 47 nm thick polymer brushes were constructed by atom transfer radical polymerization (ATRP) of poly(N-isopropylacrylamide). The resulting surfaces of the multilayered polysaccharide ultrathin films were evaluated by ellipsometry, IR imaging, in situ variable-temperature atomic force microscopy (AFM), and contact angle measurements. The morphological transformation of the freestanding polymer nanosheet bearing thermoresponsive polymer brushes was observed macroscopically through reversible structural color changes at the air-water interface. The dynamic shape change of the nanosheet was also monitored with the addition of a surfactant such as sodium n-dodecylsulfate to reduce the hydrophobicity of the surface. It was then demonstrated that the highly flexible freestanding polymer nanosheet is capable of acting as a unique platform for inducing stimuli-responsive behavior in nanomaterials.
Sigma-Aldrich
Sodium 1-dodecanesulfonate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium 1-dodecanesulfonate, Vetec, reagent grade