Merck
  • Comparative analysis of cleavable azobenzene-based affinity tags for bioorthogonal chemical proteomics.
Comparative analysis of cleavable azobenzene-based affinity tags for bioorthogonal chemical proteomics.
Chemistry & biology (2010-11-26)
Yu-Ying Yang, Markus Grammel, Anuradha S Raghavan, Guillaume Charron, Howard C Hang
ABSTRACT
The advances in bioorthogonal ligation methods have provided new opportunities for proteomic analysis of newly synthesized proteins, posttranslational modifications, and specific enzyme families using azide/alkyne-functionalized chemical reporters and activity-based probes. Efficient enrichment and elution of azide/alkyne-labeled proteins with selectively cleavable affinity tags are essential for protein identification and quantification applications. Here, we report the synthesis and comparative analysis of Na₂S₂O₄-cleavable azobenzene-based affinity tags for bioorthogonal chemical proteomics. We demonstrated that ortho-hydroxyl substituent is required for efficient azobenzene-bond cleavage and show that these cleavable affinity tags can be used to identify newly synthesized proteins in bacteria targeted by amino acid chemical reporters as well as their sites of modification on endogenously expressed proteins. The azobenzene-based affinity tags are compatible with in-gel, in-solution, and on-bead enrichment strategies and should afford useful tools for diverse bioorthogonal proteomic applications.
Sigma-Aldrich
L-Norleucine, ≥98% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Norleucine, suitable for amino acid analysis, BioReagent
Pricing and availability is not currently available.