Merck
  • Polyamide-scorpion cyclam lexitropsins selectively bind AT-rich DNA independently of the nature of the coordinated metal.
Polyamide-scorpion cyclam lexitropsins selectively bind AT-rich DNA independently of the nature of the coordinated metal.
PloS one (2011-05-17)
Anthony T S Lo, Noeris K Salam, David E Hibbs, Peter J Rutledge, Matthew H Todd
ABSTRACT
Cyclam was attached to 1-, 2- and 3-pyrrole lexitropsins for the first time through a synthetically facile copper-catalyzed "click" reaction. The corresponding copper and zinc complexes were synthesized and characterized. The ligand and its complexes bound AT-rich DNA selectively over GC-rich DNA, and the thermodynamic profile of the binding was evaluated by isothermal titration calorimetry. The metal, encapsulated in a scorpion azamacrocyclic complex, did not affect the binding, which was dominated by the organic tail.
Sigma-Aldrich
1,4,8,11-Tetraazacyclotetradecane, 98% (GC)