Merck
  • Soil and glass surface photodegradation of etofenprox under simulated california rice growing conditions.
Soil and glass surface photodegradation of etofenprox under simulated california rice growing conditions.
Journal of agricultural and food chemistry (2011-06-17)
Martice Vasquez, Thomas Cahill, Ronald Tjeerdema
ABSTRACT
Photolysis is an important degradation process to consider when evaluating a pesticide's persistence in a rice field environment. To simulate both nonflooded and flooded California rice field conditions, the photolytic degradation of etofenprox, an ether pyrethroid, was characterized on an air-dried rice soil and a flooded rice soil surface by determination of its half-life (t(1/2)), dissipation rate constant (k) and identification and quantitation of degradation products using LC/MS/MS. Photodegradation was also characterized on a glass surface alone to rule out confounding soil factors. Measured photolytic dissipation rates were used as input parameters into a multimedia environmental fate model to predict etofenprox persistence in a rice field environment. Photolytic degradation proceeded at a faster rate (0.23/day, t(1/2) = 3.0 days) on the flooded soil surface compared to the air-dried surface (0.039/day, t(1/2) = 18 days). Etofenprox degradation occurred relatively quickly on the glass surface (3.1/day, t(1/2) = 0.23 days or 5.5 h) compared to both flooded and air-dried soil layers. Oxidation of the ether moiety to the ester was the major product on all surfaces (max % yield range = 0.2 ± 0.1% to 9.3 ± 2.3%). The hydroxylation product at the 4' position of the phenoxy phenyl ring was detected on all surfaces (max % yield range = 0.2 ± 0.1% to 4.1 ± 1.0%). The air-dried soil surface did not contain detectable residues of the ester cleavage product, whereas it was quantitated on the flooded soil (max % yield = 0.6 ± 0.3%) and glass surface (max % yield = 3.6 ± 0.6%). Dissipation of the insecticide in dark controls was significantly different (p < 0.05) compared to the light-exposed surfaces indicating that degradation was by photolysis. Laboratory studies and fate model predictions suggest photolysis will be an important process in the overall degradation of etofenprox in a rice field environment.