Merck
  • Furan carcinogenicity: DNA binding and genotoxicity of furan in rats in vivo.
Furan carcinogenicity: DNA binding and genotoxicity of furan in rats in vivo.
Molecular nutrition & food research (2012-08-07)
Carolin Neuwirth, Pasquale Mosesso, Gaetano Pepe, Mario Fiore, Mike Malfatti, Ken Turteltaub, Wolfgang Dekant, Angela Mally
ABSTRACT
Furan is a potent hepatotoxicant and liver carcinogen in rodents. However, short-term tests for genotoxicity of furan are inconclusive. The aim of this study was to assess the potential of furan to covalently bind to DNA, and to assess furan genotoxicity in rats in vivo. Accelerator mass spectrometry was used to determine the (14) C-content in DNA following administration of [3,4-(14) C]-furan (0.1 and 2.0 mg/kg bw) to F344 rats. DNA damage, micronuclei, chromosomal aberrations, and sister chromatid exchanges were analyzed in F344 rats treated with furan for up to 28 days. The (14) C-content in liver DNA was significantly increased in a dose-dependent manner, with mean concentrations of 7.9 ± 3.5 amol (14) C/μg DNA and 153.3 ± 100.2 amol (14) C/μg DNA, corresponding to 16.5 ± 7.4 and 325.2 ± 212.7 adducts/10(9) nucleotides at 0.1 and 2.0 mg/kg bw, respectively. There was no evidence for genotoxicity of furan in peripheral blood and bone marrow cells. However, a dose-related increase in the incidence of chromosomal aberrations in rat splenocytes and some indication of DNA damage in liver were observed. Collectively, results from this study indicate that furan may operate-at least in part-by a genotoxic mode of action.
Product Description
Supelco
Furan, analytical standard
Sigma-Aldrich
Furan, 98%
Sigma-Aldrich
Furan, ≥99%