Merck
  • Involvement of P2Y receptors in myocardial contractile activity of rats during postnatal ontogeny.
Involvement of P2Y receptors in myocardial contractile activity of rats during postnatal ontogeny.
Bulletin of experimental biology and medicine (2012-07-18)
T A Anikina, I N Anisimova, F G Sitdikov
ABSTRACT
We studied the effect of uridine 5'-triphosphate in concentrations of 10(-10)-10(-6) M on myocardial contractile activity in 7-100-day-old rats. Analysis of isometric contraction of myocardial strips showed that uridine 5'-triphosphate reduced the strength of myocardial contraction in rats of all age groups. In 21- and 100-day-old rat pups, exogenous uridine 5'-triphosphate produced a stronger inhibitory effect than in 7-day-old animals. The negative inotropic effect of UTP was abolished under conditions of P2Y(4) purinoceptor blockade with reagent blue-2. These data indicate that the effect of UTP on the myocardium is realized via P2Y(4) purinoceptors.