- Tandem Hass-Bender/Henry reaction for the synthesis of dimethylnitro alcohols from benzylic halides.
Tandem Hass-Bender/Henry reaction for the synthesis of dimethylnitro alcohols from benzylic halides.
Tetrahedron Letters (2005)
Klein TA and Schkeryantz JM.