Merck
Fatal fulminant hepatic failure due to cyproterone acetate.
Digestive diseases and sciences (1999-09-18)
G Friedman, E Lamoureux, A H Sherker
ABSTRACT
Cyproterone acetate is a normally well-tolerated drug that is used widely for the treatment of prostatic carcinoma. Liver toxicity due to its use is not well known. We describe two cases of fatal fulminant hepatitis related to the use of cyproterone acetate.
Sigma-Aldrich
Cyproterone acetate, ≥98%
Cyproterone acetate, European Pharmacopoeia (EP) Reference Standard