  • Search Results
  • Synthesis and characterization of hexagonally packed mesoporous tantalum oxide molecular sieves.
Synthesis and characterization of hexagonally packed mesoporous tantalum oxide molecular sieves.
Chemistry of Materials (1996)
Antonelli DM and Ying JY