Merck
  • n-3 PUFAs have beneficial effects on anxiety and cognition in female rats: Effects of early life stress.
n-3 PUFAs have beneficial effects on anxiety and cognition in female rats: Effects of early life stress.
Psychoneuroendocrinology (2015-05-13)
Matteo M Pusceddu, Philip Kelly, Nurbazilah Ariffin, John F Cryan, Gerard Clarke, Timothy G Dinan
ABSTRACT
Stressful life events, especially those in early life, can exert long-lasting changes in the brain, increasing vulnerability to mental illness especially in females. Omega-3 polyunsaturated fatty acids (n-3 PUFAs) play a critical role in the development and function of the central nervous system (CNS). Thus, we investigated the influence of an eicosapentaenoic acid (EPA)/docosahexaenoic acid (DHA) (80% EPA, 20% DHA) n-3 PUFAs mixture on stress-related behavioural and neurobiological responses. Sprague-Dawley female rats were subjected to an early-life stress, maternal separation (MS) procedure from postnatal days 2 to 12. Non-separated (NS) and MS rats were administered saline, EPA/DHA 0.4g/kg/day or EPA/DHA 1g/kg/day, respectively. In adulthood, EPA/DHA treated animals had a dose dependent reduction in anxiety in NS rats. Furthermore, cognitive performance in the novel object recognition task (NOR) was improved by EPA/DHA treatment in NS animals only. EPA/DHA 1g/kg/day decreased behavioural despair in the forced swim test. Notably, EPA/DHA high dose increased the translocation of GRs into the nucleus of NS rat hippocampus. However, the levels of mBDNF remained unchanged in all the experimental groups. The corticosterone response to an acute stress was blunted in MS rats and this was further attenuated by pre-treatment with EPA/DHA. Immune response and monoamine neurotransmission were significantly altered by early-life stress. In conclusion, our study supports the view that n-3 PUFAs are beneficial in neurodevelopmentally normal animals but have little positive benefit in animals exposed to early life stress.
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, AR, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, LR, ≥99%
Sigma-Aldrich
Methanol, low water for titration
Sigma-Aldrich
Sodium hydroxide solution, 0.1 N
Sigma-Aldrich
Sodium hydroxide, LR, flakes, ≥96%
Sigma-Aldrich
Sodium hydroxide, AR, pellets, ≥98.5%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)