Merck
  • Autosomal recessive phosphoglucomutase 3 (PGM3) mutations link glycosylation defects to atopy, immune deficiency, autoimmunity, and neurocognitive impairment.
Autosomal recessive phosphoglucomutase 3 (PGM3) mutations link glycosylation defects to atopy, immune deficiency, autoimmunity, and neurocognitive impairment.
The Journal of allergy and clinical immunology (2014-03-05)
Yu Zhang, Xiaomin Yu, Mie Ichikawa, Jonathan J Lyons, Shrimati Datta, Ian T Lamborn, Huie Jing, Emily S Kim, Matthew Biancalana, Lynne A Wolfe, Thomas DiMaggio, Helen F Matthews, Sarah M Kranick, Kelly D Stone, Steven M Holland, Daniel S Reich, Jason D Hughes, Huseyin Mehmet, Joshua McElwee, Alexandra F Freeman, Hudson H Freeze, Helen C Su, Joshua D Milner
ABSTRACT
Identifying genetic syndromes that lead to significant atopic disease can open new pathways for investigation and intervention in allergy. We sought to define a genetic syndrome of severe atopy, increased serum IgE levels, immune deficiency, autoimmunity, and motor and neurocognitive impairment. Eight patients from 2 families with similar syndromic features were studied. Thorough clinical evaluations, including brain magnetic resonance imaging and sensory evoked potentials, were performed. Peripheral lymphocyte flow cytometry, antibody responses, and T-cell cytokine production were measured. Whole-exome sequencing was performed to identify disease-causing mutations. Immunoblotting, quantitative RT-PCR, enzymatic assays, nucleotide sugar, and sugar phosphate analyses, along with matrix-assisted laser desorption/ionization-time-of-flight mass spectrometry of glycans, were used to determine the molecular consequences of the mutations. Marked atopy and autoimmunity were associated with increased T(H)2 and T(H)17 cytokine production by CD4(+) T cells. Bacterial and viral infection susceptibility were noted along with T-cell lymphopenia, particularly of CD8(+) T cells, and reduced memory B-cell numbers. Apparent brain hypomyelination resulted in markedly delayed evoked potentials and likely contributed to neurologic abnormalities. Disease segregated with novel autosomal recessive mutations in a single gene, phosphoglucomutase 3 (PGM3). Although PGM3 protein expression was variably diminished, impaired function was demonstrated by decreased enzyme activity and reduced uridine diphosphate-N-acetyl-D-glucosamine, along with decreased O- and N-linked protein glycosylation in patients' cells. These results define a new congenital disorder of glycosylation. Autosomal recessive hypomorphic PGM3 mutations underlie a disorder of severe atopy, immune deficiency, autoimmunity, intellectual disability, and hypomyelination.
Sigma-Aldrich
Monoclonal Anti-PGM3 antibody produced in mouse, clone 1E2-1B12, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.