Merck
  • P68 RNA helicase (DDX5) alters activity of cis- and trans-acting factors of the alternative splicing of H-Ras.
P68 RNA helicase (DDX5) alters activity of cis- and trans-acting factors of the alternative splicing of H-Ras.
PloS one (2008-08-14)
Maria Camats, Sonia Guil, Mariette Kokolo, Montse Bach-Elias
ABSTRACT
H-Ras pre-mRNA undergoes an alternative splicing process to render two proteins, namely p21 H-Ras and p19 H-Ras, due to either the exclusion or inclusion of the alternative intron D exon (IDX), respectively. p68 RNA helicase (p68) is known to reduce IDX inclusion. Here we show that p68 unwinds the stem-loop IDX-rasISS1 structure and prevents binding of hnRNP H to IDX-rasISS1. We also found that p68 alters the dynamic localization of SC35, a splicing factor that promotes IDX inclusion. The knockdown of hnRNP A1, FUS/TLS and hnRNP H resulted in upregulation of the expression of the gene encoding the SC35-binding protein, SFRS2IP. Finally, FUS/TLS was observed to upregulate p19 expression and to stimulate IDX inclusion, and in vivo RNAi-mediated depletion of hnRNP H decreased p19 H-Ras abundance. Taken together, p68 is shown to be an essential player in the regulation of H-Ras expression as well as in a vital transduction signal pathway tied to cell proliferation and many cancer processes.