Merck
  • MicroRNA-744 promotes prostate cancer progression through aberrantly activating Wnt/β-catenin signaling.
MicroRNA-744 promotes prostate cancer progression through aberrantly activating Wnt/β-catenin signaling.
Oncotarget (2017-01-21)
Han Guan, Chunhui Liu, Fang Fang, Yeqing Huang, Tao Tao, Zhixin Ling, Zonghao You, Xu Han, Shuqiu Chen, Bin Xu, Ming Chen
ABSTRACT
Accumulated evidence indicate that miR-744 functions as either tumor suppressor or oncogene in the progression of a variety of tumors, with a tumor type-specific way. However, little is known about how miR-744 impacts on the tumorigenesis of human prostate cancer. In this study, employing the analyses of microarray, qRT-PCR and re-analysis of MSKCC data, we found that CRPC tissues expressed much more miR-744 than ADPC tissues did, and the expression level of miR-744 was inversely associated with survival of CRPC patients. In vitro studies revealed that miR-744 promotes PCa cells proliferation, enhances migration, invasion; in vivo results demonstrated that silencing of miR-744 mediated by shRNA dramatically reduces PCa xenograft tumor growth. Importantly, through human gene expression array, pathway enrichment analysis and Western blot, we identified that miR-744 dramatically activated Wnt/β-catenin pathway by targeting multiple negative regulators of Wnt/β-catenin signaling, including SFRP1, GSK3β, TLE3 and NKD1. At molecular level, we further defined that NKD1 is a major functional target of miR-744. Our findings indicate that miR-744 acts as one of oncogenic factor in the progression of CRPC by recruiting a mechanism of aberrant activation of Wnt/β-catenin signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting human NKD1