Merck
  • Anatomic and Physiologic Heterogeneity of Subgroup-A Auditory Sensory Neurons in Fruit Flies.
Anatomic and Physiologic Heterogeneity of Subgroup-A Auditory Sensory Neurons in Fruit Flies.
Frontiers in neural circuits (2017-07-14)
Yuki Ishikawa, Natsuki Okamoto, Mizuki Nakamura, Hyunsoo Kim, Azusa Kamikouchi
ABSTRACT
The antennal ear of the fruit fly detects acoustic signals in intraspecific communication, such as the courtship song and agonistic sounds. Among the five subgroups of mechanosensory neurons in the fly ear, subgroup-A neurons respond maximally to vibrations over a wide frequency range between 100 and 1,200 Hz. The functional organization of the neural circuit comprised of subgroup-A neurons, however, remains largely unknown. In the present study, we used 11
Product Description
Supelco
Serotonin, analytical standard