- A Functional Na
A Functional Na
Molecular pharmacology (2017-06-25)
Ramkumar Rajamani, Sophie Wu, Iyoncy Rodrigo, Mian Gao, Simon Low, Lisa Megson, David Wensel, Rick L Pieschl, Debra J Post-Munson, John Watson, David R Langley, Michael K Ahlijanian, Linda J Bristow, James Herrington
PMID28645932
ABSTRACT
The Na
MATERIALS