Merck
  • Branched-chain ketoacids secreted by glioblastoma cells via MCT1 modulate macrophage phenotype.
Branched-chain ketoacids secreted by glioblastoma cells via MCT1 modulate macrophage phenotype.
EMBO reports (2017-10-27)
Lidia Santos Silva, Gernot Poschet, Yannic Nonnenmacher, Holger M Becker, Sean Sapcariu, Ann-Christin Gaupel, Magdalena Schlotter, Yonghe Wu, Niclas Kneisel, Martina Seiffert, Rüdiger Hell, Karsten Hiller, Peter Lichter, Bernhard Radlwimmer
ABSTRACT
Elevated amino acid catabolism is common to many cancers. Here, we show that glioblastoma are excreting large amounts of branched-chain ketoacids (BCKAs), metabolites of branched-chain amino acid (BCAA) catabolism. We show that efflux of BCKAs, as well as pyruvate, is mediated by the monocarboxylate transporter 1 (MCT1) in glioblastoma. MCT1 locates in close proximity to BCKA-generating branched-chain amino acid transaminase 1, suggesting possible functional interaction of the proteins. Using in vitro models, we demonstrate that tumor-excreted BCKAs can be taken up and re-aminated to BCAAs by tumor-associated macrophages. Furthermore, exposure to BCKAs reduced the phagocytic activity of macrophages. This study provides further evidence for the eminent role of BCAA catabolism in glioblastoma by demonstrating that tumor-excreted BCKAs might have a direct role in tumor immune suppression. Our data further suggest that the anti-proliferative effects of MCT1 knockdown observed by others might be related to the blocked excretion of BCKAs.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-(−)-Glucose, ≥99%