Merck
  • A frameshift mutation in exon 2 of the phenylalanine hydroxylase gene linked to RFLP haplotype 1.
A frameshift mutation in exon 2 of the phenylalanine hydroxylase gene linked to RFLP haplotype 1.
Human genetics (1991-10-01)
A Eigel, B Dworniczak, L Kalaydjieva, J Horst
ABSTRACT
A deletion of a single base in codon 55 (exon 2) of the phenylalanine hydroxylase (PAH) gene has been identified by direct DNA sequencing of 94 phenylketonuria (PKU) chromosomes. This mutation alters the reading frame so that a stop signal (TAA) is generated in codon 60 of the PAH gene. Haplotype analysis revealed that all PKU alleles showing the codon 55 frameshift mutation exhibited haplotype 1. In our panel of DNA probes 13% of all mutant haplotype 1 alleles carry this particular mutation. Patients who were compound heterozygotes for this deletion and R408W in exon 12, or the splice mutation in intron 12, were affected by severe PKU. Thus, the clinical data provide additional evidence that haplotype 1 PKU alleles carry molecular defects which confer a null phenotype. In addition, we were able to show that the newly detected mutation occurs on alleles of different ethnic background.