Chromolith® WidePore 300 HPLC Columns for Bioapplications
Biotherapeutics, for example bio-engineered drugs and peptide therapeutics, represent the promise of new medical treatments for the future. Production costs have been falling leading to an extremely high demand for suitable analytical methods for process monitoring and quality control of these biomolecules. HPLC is the preferred method of analysis, and it is therefore important to use the right column for these larger molecules.
Accurate analysis of proteins, antibodies and large peptides requires columns with good permeability, along with better mass transfer and selectivity. In order for size-exclusion not to influence the separation, the pore size should be approximately ten-times larger than the molecule being analyzed.
Chromolith columns have shown great potential and superiority in comparison to standard silica. In contrast to conventional packed-particle columns, wide pore (300 Å) monolithic silica columns are made of a single continuous-bed rod of high purity porous silica that is then bonded with C18, C8, C4, Epoxy and Protein A depending on the use of the column.
In addition, Chromolith® and Chromolith® HR columns are available for separation of small molecules with different column modifications
Monolithic columns remove backpressure as the primary consideration in method development and allow:
With no individual particles to shift or break, column performance is consistent over a much longer lifetime, making them ideal for relatively ”dirty or matrix rich” sample analysis. High permeability also makes them very forgiving of less rigorously prepared samples, in addition to making it easier to aggressively flush out for reequilibration.
Chromolith® WP 300 for Biomolecule Separation |
---|