Merck
HomeCancer ResearchStable Isotopes in PET (Positron Emission Tomography)
Stable Isotopes in PET (Positron Emission Tomography)
Positron Emission Tomography
Positron Emission Tomography
Labeled isotopes used as target materials in imaging have provided insights into the various mechanisms of human physiology and have lead to treatments for diseases. Water-18O is used for the production of 18F radionuclides, such as 18FDG (Fluoro-deoxyglucose). 18FDG used in PET and with other imaging technologies, such as CT/SPECT, has led to the diagnosis and treatment of a wide range of diseases, including lung, liver, and brain tumors, epilepsy, and Alzheimer’s.
ISOTEC offers materials for the production of 18F, 11C, and 15O and also offers the complete line of reagents needed for the production of 18FDG.