Product Name
Simeton PESTANAL®, analytical standard
Product Number
32072
Product Brand
SIAL
CAS Number
673-04-1
Molecular Weight
197.24
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | White to Off White |
| APPEARANCE (FORM) | Powder or Crystals |
| PURITY (GC AREA %) | ≥ 98.0 % |
| MELTING POINT | 90 - 95 C |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 36 MONTHS |