Product Name
Eugenol PESTANAL®, analytical standard
Product Number
35995
Product Brand
SIAL
CAS Number
97-53-0
Molecular Weight
164.20
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | Colorless to Yellow |
| APPEARANCE (FORM) | Liquid |
| PURITY (GC AREA %) | ≥ 98.0 % |
| REFRACTIVE INDEX N20/D | 1.5395 - 1.5435 |
| WATER | ≤ 1.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| EXPIRY PERIOD | 60 MONTHS |