Skip to Content
Merck

BL3H160BBDE3

BLD Pharmatech

4,6-Diaminopyrimidine-2-thiol

Synonym(s):

4,6-diamino-2-pyrimidinethiol

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C4H6N4S1
CAS Number:
Molecular Weight:
142.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C4H6N4S/c5-2-1-3(6)8-4(9)7-2/h1H,(H5,5,6,7,8,9)

Inchi Key

QCAWOHUJKPKOMD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: Slightly pale yellow - Yellow

Melting Point

>280C

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service