Iupac Name
1-(dimethylamino)-2-propanol
Inchi Code
1S/C5H13NO/c1-5(7)4-6(2)3/h5,7H,4H2,1-3H3
InChI key
NCXUNZWLEYGQAH-UHFFFAOYSA-N
storage temp.
Ambient Storage
Purity
95.00%
Physical Form
Liquid
Product Link
https://fluorochem.co.uk/product/F092070
Signal Word
Danger
Hazard Statements
MSDS
https://sds.fluorochem.co.uk/eng/sds_F092070_en.pdf
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service
