Iupac Name
acetic acid hydrochloride
Inchi Code
1S/C2H4O2.ClH/c1-2(3)4;/h1H3,(H,3,4);1H
InChI key
WJGAPUXHSQQWQF-UHFFFAOYSA-N
storage temp.
Ambient Storage
Physical Form
Liquid
Product Link
https://fluorochem.co.uk/product/F094031
Signal Word
Warning
Hazard Statements
MSDS
https://sds.fluorochem.co.uk/eng/sds_F094031_en.pdf
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service