Skip to Content
Merck

FLUH99C80E58

Fluorochem (Preferred Partner)

Phosphomolybdic acid hydrate

Synonym(s):

molybdenum(III) tritriacontahydroxide phosphate trihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/12Mo.H3O4P.36H2O/c;;;;;;;;;;;;1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;;;;;;;;;;;;(H3,1,2,3,4);36*1H2/q12*+3;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-36

Inchi Key

COGITEDADTZLKP-UHFFFAOYSA-A

Storage temp.

Ambient Storage

Country of Origin

GB

Physical Form

Solid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service