Skip to Content
Merck

FLUH99C88C5F

Fluorochem (Preferred Partner)

4,6-Diaminopyrimidine-2-thiol

Synonym(s):

4,6-diamino-2-pyrimidinethiol

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C4H6N4S1
CAS Number:
Molecular Weight:
142.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C4H6N4S/c5-2-1-3(6)8-4(9)7-2/h1H,(H5,5,6,7,8,9)

Inchi Key

QCAWOHUJKPKOMD-UHFFFAOYSA-N

Purity

98%

Storage temp.

Ambient Storage

Country of Origin

GB

Physical Form

Liquid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service