Merck
All Photos(1)

Documents

CRM020

Supelco

Trace Metals - Sandy Loam 2

certified reference material, pkg of 50 g

Sign Into View Organizational & Contract Pricing

NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM020-050

technique(s)

AAS: suitable
ICP: suitable

pH

2.96

application(s)

environmental

format

matrix material

storage temp.

2-30°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CRM043CRM025CRM021
Supelco

CRM020

Trace Metals - Sandy Loam 2

Supelco

CRM043

Trace Metals - Sandy Loam 6

Supelco

CRM025

Trace Metals - Sandy Loam 8

Supelco

CRM021

Trace Metals - Sandy Loam 3

application(s)

environmental

application(s)

environmental

application(s)

environmental

application(s)

environmental

format

matrix material

format

matrix material

format

matrix material

format

matrix material

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

packaging

pkg of 50 g

packaging

pkg of 50 g

packaging

pkg of 50 g

packaging

pkg of 100 g

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis. Download your certificate of analysis at: http://www.sigma-aldrich.com.

Other Notes

The following elements are not certified and are included for information only:
  • K, Na, Sr
Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Aluminum
    Antimony
    Arsenic
    Barium
    Cadmium
    Calcium
    Chromium
    Cobalt
    Copper
    Iron
    Lead
    Magnesium
    Manganese
    Mercury
    Nickel
    Selenium
    Silver
    Sodium
    Strontium
    Thallium
    Vanadium
    Zinc
    See All (22)

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Flam. Sol. 2 - Repr. 1A - Self-heat. 2 - Skin Sens. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service