Skip to Content
Merck

NIST3156

Tellurium (Te) Standard Solution

NIST®SRM® 3156

Synonym(s):

Tellurium chloride

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
12352200
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

solution

General description

This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of tellurium. SRM 3156_cert SRM 3156 _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service