Merck
All Photos(1)

SRE0102

Sigma-Aldrich

Carbonate Bicarbonate Buffer

pH 9.4 - 9.8 after reconstitution

form

powder

Quality Level

quality

pH 9.4 - 9.8 after reconstitution

shipped in

wet ice

storage temp.

2-8°C

Application

Na-carbonate (carb-bicarb) buffer for use in protein and antibody coating procedures. Intended applications include use as a coating buffer for protein-coating procedures on microtiter plates and plastic tubes for radioimmunoassay (RIA) and enzyme immunoassay (EIA) techniques.

Physical form

Product is a preformulated powder to be dissolved for use. Prepare the Single Use Disposable Solution (SUDS) by dissolving the powder in 1L of purified water for a 1× working solution of 0.05 M sodium carbonate-bicarbonate buffer with pH 9.6 at 25 °C. Approximately 70:30 bicarb:carb ratio.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service