Iupac Name
3-isobutyl-1-methyl-3,9-dihydro-1H-purine-2,6-dione
Inchi Code
1S/C10H14N4O2/c1-6(2)4-14-8-7(11-5-12-8)9(15)13(3)10(14)16/h5-6H,4H2,1-3H3,(H,11,12)
InChI key
APIXJSLKIYYUKG-UHFFFAOYSA-N
storage temp.
-80/-20
Purity
100%
Country of Origin
CN
Shipping Temperature
4
Physical Form
solid
Product Link
https://www.targetmol.com/compound/IBMX
MSDS
https://www.targetmol.com/compound/IBMX
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service