Iupac Name
disodium (2S,5R,6R)-3,3-dimethyl-6-{[(2R)-3-oxido-3-oxo-2-(2-thienyl)propanoyl]amino}-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Inchi Code
1S/C15H16N2O6S2.2Na/c1-15(2)9(14(22)23)17-11(19)8(12(17)25-15)16-10(18)7(13(20)21)6-4-3-5-24-6;;/h3-5,7-9,12H,1-2H3,(H,16,18)(H,20,21)(H,22,23);;/q;2*+1/p-2/t7-,8-,9+,12-;;/m1../s1
InChI key
JHEIXNWVYNQSRN-QBGWIPKPSA-L
storage temp.
-80/-20
Purity
99.01%
Country of Origin
CN
Shipping Temperature
4
Physical Form
solid
Product Link
https://www.targetmol.com/compound/Ticarcillin disodium
MSDS
https://www.targetmol.com/compound/Ticarcillin disodium
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service