Merck
L-Asparagine monohydrate EMPROVE® EXPERT, Ph. Eur., ChP

L-Asparagine monohydrate

L-Asparagine monohydrate EMPROVE® EXPERT, Ph. Eur., ChP
Synonym(s):
L-Asparagine monohydrate
Empirical Formula (Hill Notation):
C4H8N2O3 · xH2O
CAS Number:
Molecular Weight:
132.12 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Optical Isomer
Showing 1-2 of 2