Merck
Potassium Phosphate, Dibasic, Trihydrate, Molecular Biology Grade

Potassium phosphate dibasic trihydrate

Potassium Phosphate, Dibasic, Trihydrate, Molecular Biology Grade
Synonym(s):
Dipotassium hydrogen phosphate trihydrate, Dipotassium phosphate, di-Potassium hydrogen phosphate trihydrate, Potassium Phosphate, Dibasic, Trihydrate, Molecular Biology Grade
Linear Formula:
K2HPO4 · 3H2O
CAS Number:
Molecular Weight:
228.22

Products

Product Category
Brand
Color
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Technique