Skip to Content
Merck
trans-1,2-Cyclohexanediol 98%

trans-1,2-Cyclohexanediol

trans-1,2-Cyclohexanediol 98%
Synonym(s):
trans-1,2-Cyclohexanediol
Linear Formula:
C6H10-1,2-(OH)2
CAS Number:
Molecular Weight:
116.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing