NL
EN
Applications
Products
Services
Support
Home
Search Results
Mizoroki-Heck-type reaction mediated by potassium tert-butoxide
Mizoroki-Heck-type reaction mediated by potassium tert-butoxide
Angewandte Chemie (International ed. in English) (2011)
Shirakawa E, et al.