Merck
  • Long-acting beta2-agonists versus theophylline for maintenance treatment of asthma.
Long-acting beta2-agonists versus theophylline for maintenance treatment of asthma.
The Cochrane database of systematic reviews (2007-07-20)
A K H Tee, M S Koh, P G Gibson, T J Lasserson, A J Wilson, L B Irving
ABSTRACT
Theophylline and long acting beta-2 agonists are bronchodilators used for the management of persistent asthma symptoms, especially nocturnal asthma. They represent different classes of drug with differing side-effect profiles. To assess the comparative efficacy, safety and side-effects of long-acting beta-2 agonists and theophylline in the maintenance treatment of adults and adolescents with asthma. We searched the Cochrane Airways Group trials register and reference lists of articles. We also contacted authors of identified RCTs for other relevant published and unpublished studies and pharmaceutical manufacturers. Most recent search: November 2006. All included studies were RCTs involving adults and children with clinical evidence of asthma. These studies must have compared oral sustained release and/or dose adjusted theophylline with an inhaled long-acting beta-2 agonist. In original review, two reviewers independently assessed trial quality and extracted data, similarly in this update two reviewers undertook this. Study authors were contacted for additional information. Thirteen studies with a total of 1344 participants met the inclusion criteria of the review. They were of varying quality. There was no significant difference between salmeterol and theophylline in FEV(1) predicted (6.5%; 95% CI -0.84 to 13.83). However, salmeterol treatment led to significantly better morning PEF (mean difference 16.71 L/min, 95% CI 8.91 to 24.51) and evening PEF (mean difference 15.58 L/min, 95% CI 8.33 to 22.83). Salmeterol also reduced the use of rescue medication. Formoterol, used in two studies was reported to be as effective as theophylline. Bitolterol, used in only one study, was reported to be less effective than theophylline. Participants taking salmeterol experienced fewer adverse events than those using theophylline (Parallel studies: Relative Risk 0.44; 95% CI 0.30 to 0.63, Risk Difference -0.11; 95% CI -0.16 to -0.07, Numbers Needed to Treat (NNT) 9; 95% CI 6 to 14). Significant reductions were reported for central nervous system adverse events (Relative Risk 0.50; 95% CI 0.29 to 0.86, Risk Difference -0.07; 95% CI -0.12 to -0.02, NNT 14; 95% CI 8 to 50) and gastrointestinal adverse events (Relative Risk 0.30; 95% CI 0.17 to 0.55, Risk Difference -0.11; 95% CI -0.16 to -0.06, NNT 9; 95% CI 6 to 16). Long-acting beta-2 agonists, particularly salmeterol, are more effective than theophylline in improving morning and evening PEF, but are not significantly different in their effect on FEV1. There is evidence of decreased daytime and nighttime short-acting beta-2 agonist requirement with salmeterol. Fewer adverse events occurred in participants using long-acting beta-2 agonists (salmeterol and formoterol) as compared to theophylline.
Product Description
USP
Theophylline, certified reference material, United States Pharmacopeia (USP) Reference Standard
Theophylline, European Pharmacopoeia (EP) Reference Standard
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder