Merck
HPLC Analysis of Nucleosides on Ascentis® Si (HILIC Mode)
HPLC Analysis of Nucleosides on Ascentis® Si (HILIC Mode) application for HPLC
Materials
related product
Product No.
Description
Pricing
Ascentis® Si HPLC Column
5 μm particle size, L × I.D. 10 cm × 2.1 mm
CONDITIONS
column
Ascentis Si, 10 cm x 2.1 mm I.D., 5 μm particles (581508-U)
mobile phase
[A] 150 mM ammonium formate (pH 3.0, with formic acid): [B] acetonitrile; (10:90, A:B)
flow rate
0.2 mL/min
column temp.
35 °C
detector
UV, 270 nm
injection
1 μL
sample
as indicated in mobile phase
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany