Merck
UHPLC/MS Analysis of 8-Hydroxy-2′-Deoxyguanosine and Related Compounds on Titan Silica
UHPLC/MS Analysis of 8-Hydroxy-2′-Deoxyguanosine and Related Compounds on Titan Silica application for UHPLC-MS
Materials
related product
Product No.
Description
Pricing
8-Hydroxy-2′-deoxyguanosine
≥98% (TLC)
2′-Deoxyguanosine monohydrate
99-100%
Guanosine
≥98%
CONDITIONS
column
Titan Silica, 10 cm x 3.0 mm I.D., 1.9 μm particles (581536-U)
mobile phase
[A] acetonitrile; [B] 100 mM ammonium formate, pH 3.0 with formic acid; (90:10, A:B)
flow rate
0.6 mL/min
pressure
2060 psi (142 bar)
column temp.
35 °C
detector
MS ESI+, combined SIR, 268, 284 m/z
injection
2 μL
sample
1 μg/mL in 99:1, acetonitrile:water
Description
Analysis Note
The rapid, efficient separation of these highly polar compounds on a Titan Silica column in HILIC mode is shown here.
Legal Information
null