Merck
HomePhotometry & ReflectometryAnalytical Method: Tartaric Acid in Water
Analytical Method: Tartaric Acid in Water
Reagents:
Cat.No. 101512 Silver nitrate for analysis
Cat.No. 100063 Acetic acid 100 % for analysis
Cat.No. 116754 Water for analysis
Cat.No. 109137 Sodium hydroxide solution 1 mol/l
Cat.No. 106268 Sodium acetate for analysis
Clarocarbon G
Ammonium vanadate for analysis
Reagent 1:
Dissolve 1.7 g silver nitrate in 20 ml water for analysis, add 30 ml acetic acid and make up to mark with water for analysis in a 100-ml volumetric flask.
Reagent 2:
Clarocarbon G
Reagent 3:
Dissolve 1.0 g ammonium vanadate in 15 ml sodium hydroxide solution 1 mol/l. Add 5.4 g sodium acetate and make up to mark with water for analysis in a 100-mL volumetric flask.
Measurement range:
0.20 - 3.50 mg/L tartaric acid at cell length of 10 mm and 445/446 nM
Analysis:
Mix 3.0 ml sample with 10 mL reagent 1. Add 0.5 g reagent 2 and shake. Add 10 ml reagent 3 and mix and filter off the carbon using a folded filter. Measure the clear particle-free filtrate against water at 445 nm (in the photometer SQ 118 at 446 nM).
Literature:
A. Schmitt: Aktuelle Weinanalytik, 2. Auflage 1983. Verlag Heller Chemie- und Verwaltungs-gesellschaft mbH. Schwäbisch-Hall
Parameter list for programming the photometer Spectroquant® NOVA 60, NOVA 400, Pharo 100, and Pharo 300
Calculation when measuring the absorbance value using the above NOVA photometers without programming the method in the memory of the instrument:
Parameter list for programming the photometer SQ 118
Calculation when measuring the absorbance value using the photometer SQ118 without programming the method in the memory of the instrument: