Build with Better Bases
Reliable oligonucleotide reagents from a supplier with greater capacity, custom capabilities and packaging, QA & documentation, and complete support of your most demanding project.
Reagents for Oligonucleotide Synthesis
The Proligo® reagents portfolio offers a full range of phosphoramidites, solid supports, and liquid reagents for oligonucleotide synthesis that include the following features:
  • High-quality standard and specialized products for the complete workflow
  • Flexible and customizable production and packaging options
Proligo® Catalog
Featuring all product and technical information associated with the Proligo® Reagents range.
Request Yours
Workflow
Synthesis Supports
Synthesis Supports
Controlled Pore Glass (CPG) has proven to be the most accepted supports for the chemical synthesis of DNA and RNA. Features include high surface area, efficient mass transfer and chemical inertness.
Detritylation & Activation
Coupling takes place once the DMT groups are removed through the detritylation mechanism utilizing TCA.
Activator solutions are mixed with solutions of phosphoramidites during the extension step.
Coupling
Phosphoramidite monomers are added through the coupling mechanism utilizing the following products:
Capping & Oxidation
Stabilization of the reactions and efficiency of the yield is achieved through oxidation and capping.
Supporting Technical Documents
Highlights
Dedicated process technology group and in-process controls that will adapt to evolving diagnostic and pharmaceutical requirements.
Quality Assurance
Dedicated process technology group and in-process controls that will adapt to evolving diagnostic and pharmaceutical requirements.
For more information about our products and services, request a quote or give us feedback on the type of information you would like to see on this page, please Contact Us.
Contact Us
For more information about our products and services, request a quote or give us feedback on the type of information you would like to see on this page, please Contact Us.
Complete packaging services from routine to custom and from milligrams to kilograms at our state-of-the-Art Manufacturing facility in Hamburg.
State-of-the-Art Production
Complete packaging services from routine to custom and from milligrams to kilograms at our state-of-the-Art Manufacturing facility in Hamburg.