Merck
All Photos(2)

5.01260

SAFC

Pyridoxol (pyridoxine)

vitamin B₆ base EMPROVE® ESSENTIAL

Synonym(s):
Pyridoxol (pyridoxine), Pyridoxine, Vitamin B6
Empirical Formula (Hill Notation):
C8H11NO3
CAS Number:
Molecular Weight:
169.18
MDL number:
EC Index Number:
200-603-0

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

potency

4000 mg/kg LD50, oral (Rat)

pH

6.5-7.0 (25 °C, 50 g/L in H2O)

mp

156-159 °C

solubility

120 g/L

bulk density

300 kg/m3

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

InChI

1S/C8H11NO3/c1-5-8(12)7(4-11)6(3-10)2-9-5/h2,10-12H,3-4H2,1H3

InChI key

LXNHXLLTXMVWPM-UHFFFAOYSA-N

General description

As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Pyridoxol (Vitamin B6) is used in clinical nutrition for treatment of vitamin B6 deficiency, anemia, mental retardation, Parkinson′s syndrome, gestational diabetes, tetanus and atherosclerosis . Pyridoxol (Vitamin B6) Emprove® Essential is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials.

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service