Merck
99100108
Opentrons® 200μL tip refill
new
Tips for AAW Workstation
Slide 1 of 1
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
41121608
Volume:
200 μL
Sterility:
sterile
Feature:
DNA free (Human)
PCR clean
RNase and DNase free
autoclavable
endotoxin free
pyrogen free
Packaging:
pkg of 20 packs of 96 each
Manufacturer/tradename:
991-00108
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene
sterility
sterile
feature
DNA free (Human)
PCR clean
RNase and DNase free
autoclavable
endotoxin free
pyrogen free
manufacturer/tradename
991-00108
packaging
pkg of 20 packs of 96 each
volume
200 μL
General description
200 µL pipette tips for the Opentrons Flex, provided sterile in racks or blister packs (refills). Tips are clear, non-conducting polypropylene tips, free of human DNA, PCR inhibitors, RNase/DNase, pyrogens, and endotoxins. Opentrons Flex Tips and Racks are autoclavable.
Legal Information
AAW is a trademark of Millipore Corporation
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Opentrons is a registered trademark of Opentrons Labworks, Inc.