Merck
LPS-Induced Acute Kidney Injury Is Mediated by Nox4-SH3YL1.
Cell reports (2020-10-22)
Jung-Yeon Yoo, Dae Ryong Cha, Borim Kim, Eun Jung An, Sae Rom Lee, Jin Joo Cha, Young Sun Kang, Jung Yeon Ghee, Jee Young Han, Yun Soo Bae
ABSTRACT
Cytosolic proteins are required for regulation of NADPH (nicotinamide adenine dinucleotide phosphate) oxidase (Nox) isozymes. Here we show that Src homology 3 (SH3) domain-containing YSC84-like 1 (SH3YL1), as a Nox4 cytosolic regulator, mediates lipopolysaccharide (LPS)-induced H2O2 generation, leading to acute kidney injury. The SH3YL1, Ysc84p/Lsb4p, Lsb3p, and plant FYVE proteins (SYLF) region and SH3 domain of SH3YL1 contribute to formation of a complex with Nox4-p22phox. Interaction of p22phox with SH3YL1 is triggered by LPS, and the complex induces H2O2 generation and pro-inflammatory cytokine expression in mouse tubular epithelial cells. After LPS injection, SH3YL1 knockout mice show lower levels of acute kidney injury biomarkers, decreased secretion of pro-inflammatory cytokines, decreased infiltration of macrophages, and reduced tubular damage compared with wild-type (WT) mice. The results strongly suggest that SH3YL1 is involved in renal failure in LPS-induced acute kidney injury (AKI) mice. We demonstrate that formation of a ternary complex of p22phox-SH3YL1-Nox4, leading to H2O2 generation, induces severe renal failure in the LPS-induced AKI model.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®
Sigma-Aldrich
Src Inhibitor-1, ≥98% (HPLC)
Sigma-Aldrich
Syk Inhibitor, The Spleen Tyrosine Kinase Inhibitor, also referenced under CAS 622387-85-3, controls the biological activity of Spleen Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.