Merck
  • MicroRNA-15b shuttled by bone marrow mesenchymal stem cell-derived extracellular vesicles binds to WWP1 and promotes osteogenic differentiation.
MicroRNA-15b shuttled by bone marrow mesenchymal stem cell-derived extracellular vesicles binds to WWP1 and promotes osteogenic differentiation.
Arthritis research & therapy (2020-11-18)
Yanhong Li, Jing Wang, Yanchao Ma, Wenjia Du, Haijun Feng, Kai Feng, Guangjie Li, Shuanke Wang
ABSTRACT
Osteogenic differentiation is an essential process for bone regeneration involving bone marrow mesenchymal stem cells (BMSCs). BMSC-secreted extracellular vesicles (EVs) enriched with microRNAs (miRs) have vital roles to play in mediating osteogenic differentiation. Therefore, this study aimed to explore the effect of BMSC-derived EVs loaded with miR-15b on osteogenic differentiation. Human BMSCs (hBMSCs) were cultured and treated with plasmids overexpressing or knocking down KLF2, WWP1, and miR-15b to define the role of derived EVs in osteogenic differentiation in vitro. The expression of osteogenic differentiation-related marker was measured by Western blot analysis. The interaction among miR-15b, WWP1, and ubiquitination of KLF2 was investigated by dual-luciferase reporter, immunoprecipitation, and GST pull-down assays. Moreover, EVs from hBMSCs transfected with miR-15b inhibitor (EV-miR-15b inhibitor) were injected into ovariectomized rats to verify the effect of miR-15b on bone loss in vivo. WWP1 was downregulated, and KLF2 was upregulated during osteogenic differentiation. After co-culture with EVs, miR-15b expression was elevated and WWP1 expression was reduced in hBMSCs. Upregulation of miR-15b or KLF2 or downregulation of WWP1 or NF-κB increased ALP activity and cell mineralization, as well as osteogenic differentiation-related marker expression in hBMSCs. Mechanistically, miR-15b targeted and inhibited WWP1, thus attenuating KLF2 degradation and inhibiting NF-κB activity. Co-culture of EVs increased the bone volume and trabecular number, but decreased bone loss in ovariectomized rats, which could be reversed after treatment with EV-miR-15b inhibitor. Collectively, BMSC-derived EVs loaded with miR-15b promoted osteogenic differentiation by impairing WWP1-mediated KLF2 ubiquitination and inactivating the NF-κB signaling pathway.
Sigma-Aldrich
Monoclonal Anti-c-Myc antibody produced in mouse, clone 9E10, ascites fluid
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
MISSION® esiRNA, targeting human WWP1